Learn More
Thermo Scientific Chemicals N-Acetyl-D-glucosamine, 98+%
N-Acetyl-D-glucosamine, CAS # 72-87-7, is the D isomer of the glucose-derived monomer found in structural polysaccharides.
£19.30 - £114.57
Chemical Identifiers
CAS | 7512-17-6 |
---|---|
Molecular Formula | C8H15NO6 |
Molecular Weight (g/mol) | 221.21 |
MDL Number | MFCD00061615 |
InChI Key | OVRNDRQMDRJTHS-WZWLWIOANA-N |
IUPAC Name | N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
SMILES | CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479983
|
Thermo Scientific Chemicals
A13047.09 |
10 g |
£19.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489983
|
Thermo Scientific Chemicals
A13047.18 |
50 g | N/A | ||||||
11438770
|
Thermo Scientific Chemicals
A13047.22 |
100 g |
£114.57
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Acetyl-D-glucosamine is used in the identification and characterization of N-acetyl-beta-D-hexoaminidase. It is also used in the treatment of osteoarthritis and inflammatory bowel disease (IBD), including ulcerative colitis and Crohn's disease. Further, it is used as a substrate in sialic acid production, cosmetics and in drug development research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• N-Acetyl-D-glucosamine is the acetylated form of glucosamine, a glucose derivative
• It has a role as a prokaryotic metabolism and monomer used in chitin, hyaluronic acid, and polymers within the cell wall
Application
• N-Acetyl-D-Glucosamine can be used for molecular binding studies; research has shown that it attaches to certain microbes and their pathogens
• It is useful for biochemical characterization studies of related glucosamines
Chemical Identifiers
7512-17-6 | |
221.21 | |
OVRNDRQMDRJTHS-WZWLWIOANA-N | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O |
C8H15NO6 | |
MFCD00061615 | |
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide |
Specifications
7512-17-6 | |
≥98% | |
MFCD00061615 | |
Hygroscopic | |
OVRNDRQMDRJTHS-WZWLWIOANA-N | |
N-[(3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide | |
221.21 |
213°C (decomposition) | |
C8H15NO6 | |
1727589 | |
Soluble in concentrated hydrochloric acid,sulfuric acid,phosphoric acid and formic acid. Insoluble in water,dilute acids,dilute alkalis,concentrated alkalis and organic solvents. | |
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@@H](O)[C@@H]1O | |
221.21 | |
N-Acetyl-D-glucosamine |
Safety and Handling
missing translation for 'einecsNumber' : 231-368-2
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only