missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-mannosamine, MedChemExpress

Product Code. 16406036
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16406036

Brand: Medchem Express HY128850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress N-Acetyl-D-mannosamine (ManNAc) is an essential precursor of N-acetylneuraminic acid (NeuAc), the specific monomer of bacterial capsular polysialic acid (PA). N-Acetyl-D-mannosamine (ManNAc) can be metabolized by GNE and GlcNAc 2-epimerase (Renin binding protein, RnBP), into ManNAc-6-phosphate and GlcNAc, respectively.

N-Acetyl-d-mannosamine (ManNAc) and its derivatives activates hypocretin (HCRT) gene expression in the orexin neurons, providing a potential model for the testing of a therapy for neural disorders.

Specifications

Chemical Name or Material N-Acetyl-D-mannosamine
Physical Form Solid
CAS 3615-17-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C8H15NO6
For Use With (Application) Neuroscience-Neuromodulation
Synonym N-Acetylmannosamine ManNAc
Solubility Information H2O : 125 mg/mL (565.07 mM; Need ultrasonic)
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](C=O)NC(C)=O
Molecular Weight (g/mol) 221.21
Formula Weight 221.21
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.