missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-DL-methionine, 99%

Catalog Number p-7026074
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
1115-47-5
C7H13NO3S
191.25
MFCD00008681
XUYPXLNMDZIRQH-UHFFFAOYNA-N
n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
6180
CSCCC(NC(C)=O)C(O)=O
This item is not returnable. View return policy
1115-47-5
C7H13NO3S
191.25
MFCD00008681
XUYPXLNMDZIRQH-UHFFFAOYNA-N
n-acetyl-dl-methionine, ac-dl-met-oh, n-acetylmethionine, acetyl-dl-methionine, dl-n-acetylmethionine, methionamine, methionine, n-acetyl, dl-methionine, n-acetyl, methionine, n-acetyl-, dl, dl-acetylmethionine
6180
CSCCC(NC(C)=O)C(O)=O

N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-methionine is used as pharmaceutical intermediate, biochemical reagent.

Solubility
Soluble in water, ethanol, ethyl acetate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 113°C to 117°C
Beilstein 1725554
Merck Index 14,96
Solubility Information Soluble in water,ethanol,ethyl acetate.
IUPAC Name 2-acetamido-4-(methylsulfanyl)butanoic acid
Formula Weight 191.25
Percent Purity 99%
Quantity 50 g
Chemical Name or Material N-Acetyl-DL-methionine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'