Learn More
Thermo Scientific Chemicals N-Acetyl-L-cysteine, 98+%
Description
A mucolytic agent for isolation of mycobacteria from sputum.N-Acetyl-L-cysteine is used in the liquefaction and decontamination of sputum during the preparation for recovery of mycobacterium, which is called as Petroff's method. It shows antiviral activity against the influenza A viruses. It is used in the treatment of interstitial lung disease to prevent disease progression. It is an antioxidant, dietary supplement and protects liver. Further, it forms complexes with palladium, thereby it dissolves in water and remove palladium from drugs. Further, it is used in the treatment of paracetamol overdose. In addition to this, it is involved in the treatment of autism, Alzheimer's disease, bipolar disorder, major depressive disorder, obsessive-compulsive disorder, schizophrenia, and specific drug addictions like cocaine and cannabis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- N-Acetyl-L-cysteine (acetylcysteine) is a precursor of the antioxidant glutathione, or GSH
- N-Acetyl-L-cysteine is cytoprotective and may prevent cancer, neurodegeneration, and other disease
Application
- N-Acetyl-L-cysteine is useful for cellular redox study because of its anti-inflammatory properties
- It may also be useful for psychiatric research and the study of neurodegeneration
- It is suitable for in vivo and in vitro experiments

Specifications
Specifications
Melting Point | 108°C to 111°C |
Assay Percent Range | ≥98% |
Beilstein | 1724426 |
Solubility Information | Soluble in water,ethanol,methanol,dimethyl sulfoxide,hot isopropyl alcohol,methyl acetate and ethyl acetate. Insoluble in chloroform and ether. |
Quantity | 100 g |
Formula Weight | 163.2 |
Chemical Name or Material | N-Acetyl-L-cysteine |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M27L020 | Certificate of Analysis | 29/01/2025 | A15409.36,A15409.14,A15409.22 |
Z17K013 | Certificate of Analysis | 23/01/2025 | A15409.14,A15409.36,A15409.22 |
M20L026 | Certificate of Analysis | 22/01/2025 | A15409.14,A15409.36,A15409.22 |
10252847 | Certificate of Analysis | 28/10/2024 | A15409.14,A15409.36,A15409.22 |
W06K031 | Certificate of Analysis | 18/09/2024 | A15409.14,A15409.36,A15409.22 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.