missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-L-glutamic acid, 99%

£22.20 - £52.60

Chemical Identifiers

CAS 1188-37-0
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.167
MDL Number MFCD00002802
InChI Key RFMMMVDNIPUKGG-YFKPBYRVSA-N
Synonym n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
PubChem CID 70914
ChEBI CHEBI:17533
IUPAC Name (2S)-2-acetamidopentanedioic acid
SMILES CC(=O)NC(CCC(=O)O)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11445657
View Documents
Thermo Scientific Chemicals
B23621.14
25 g
£22.20
25g
Please to purchase this item. Need a web account? Register with us today!
11455657
View Documents
Thermo Scientific Chemicals
B23621.22
100 g
£52.60
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-L-glutamic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water. 2.7 g/100 mL (20°C)

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Chemical Identifiers

1188-37-0
189.167
RFMMMVDNIPUKGG-YFKPBYRVSA-N
70914
(2S)-2-acetamidopentanedioic acid
C7H11NO5
MFCD00002802
n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
CHEBI:17533
CC(=O)NC(CCC(=O)O)C(=O)O

Specifications

194°C to 199°C
C7H11NO5
1727473
Soluble in water. 2.7g/100 mL (20°C)
CC(=O)NC(CCC(=O)O)C(=O)O
189.167
CHEBI:17533
−16.5° (c=1 in Water)
N-Acetyl-L-glutamic acid
1188-37-0
MFCD00002802
n-acetyl-l-glutamic acid, acetylglutamic acid, n-acetyl-l-glutamate, n-acetylglutamic acid, n-acetylglutamate, acetyl glutamic acid, acetyl-l-glutamic acid, l-glutamic acid, n-acetyl, l-n-acetylglutamic acid, ac-glu-oh
RFMMMVDNIPUKGG-YFKPBYRVSA-N
(2S)-2-acetamidopentanedioic acid
70914
189.17
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 214-708-4

missing translation for 'rtecsNumber' : LZ9725000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only