missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Acetyl-L-tyrosine, 99%
Click to view available options
Quantity:
10 g
50 g
250 g
Description
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water (25 mg/ml), and ethanol.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
N-Acetyl-L-tyrosine is involved in catecholamine production. It can be used as a cell culture media component in the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.
Solubility
Soluble in water (25 mg/ml), and ethanol.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 150°C to 151°C |
Beilstein | 2697172 |
Solubility Information | Soluble in water (25mg/ml),and ethanol. |
Formula Weight | 223.23 |
Percent Purity | 99% |
Quantity | 10 g |
Optical Rotation | +58° (c=5 in Ethanol) |
Chemical Name or Material | N-Acetyl-L-tyrosine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'