Learn More
N-Acetylethylenediamine, tech. 90%, Thermo Scientific Chemicals
£48.90 - £168.00
Chemical Identifiers
CAS | 1001-53-2 |
---|---|
Molecular Formula | C4H10N2O |
Molecular Weight (g/mol) | 102.137 |
MDL Number | MFCD00008163 |
InChI Key | DAKZISABEDGGSV-UHFFFAOYSA-N |
Synonym | n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine |
PubChem CID | 66082 |
IUPAC Name | N-(2-aminoethyl)acetamide |
SMILES | CC(=O)NCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11363027
|
Thermo Scientific Alfa Aesar
L13398.06 |
5 g |
£48.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373027
|
Thermo Scientific Alfa Aesar
L13398.14 |
25 g |
£168.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
1001-53-2 | |
102.137 | |
DAKZISABEDGGSV-UHFFFAOYSA-N | |
66082 | |
CC(=O)NCCN |
C4H10N2O | |
MFCD00008163 | |
n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine | |
N-(2-aminoethyl)acetamide |
Specifica
1001-53-2 | |
128°C (3 mmHg) | |
C4H10N2O | |
MFCD00008163 | |
1743120 | |
n-2-aminoethyl acetamide, n-acetylethylenediamine, acetamide, n-2-aminoethyl, n-2-amino-ethyl-acetamide, unii-fql33v3kzd, fql33v3kzd, acetylethylenediamine, 2-acetamidoethylamine, acetylethylene diamine, n-acetylethylendiamine | |
CC(=O)NCCN | |
102.137 | |
102.14 | |
Technical |
44°C to 50°C | |
>110°C (230°F) | |
CH3CONHCH2CH2NH2 | |
UN3259 | |
Hygroscopic | |
DAKZISABEDGGSV-UHFFFAOYSA-N | |
N-(2-aminoethyl)acetamide | |
66082 | |
90% | |
N-Acetylethylenediamine |
Sicurezza e movimentazione
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only