Learn More
N-Acetylsulfanilyl chloride, 99%, Thermo Scientific Chemicals
£19.90 - £220.00
Chemical Identifiers
CAS | 121-60-8 |
---|---|
Molecular Formula | C8H8ClNO3S |
Molecular Weight (g/mol) | 233.67 |
MDL Number | MFCD00007442 |
InChI Key | GRDXCFKBQWDAJH-UHFFFAOYSA-N |
Synonym | n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride |
PubChem CID | 8481 |
IUPAC Name | 4-acetamidobenzenesulfonyl chloride |
SMILES | CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10417740
|
Thermo Scientific Acros
150721000 |
100 g | Glass bottle |
£19.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10215440
|
Thermo Scientific Acros
150725000 |
500 g | Glass bottle |
£50.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434565
|
Thermo Scientific Acros
150720025 |
2.5 kg | Plastic Bottle |
£220.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
121-60-8 | |
233.67 | |
GRDXCFKBQWDAJH-UHFFFAOYSA-N | |
8481 | |
CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O |
C8H8ClNO3S | |
MFCD00007442 | |
n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride | |
4-acetamidobenzenesulfonyl chloride |
Specifications
121-60-8 | |
100.0 | |
Beige-Cream to White | |
Authentic | |
C8H8ClNO3S | |
MFCD00007442 | |
01,3 | |
n-acetylsulfanilyl chloride, 4-acetamidobenzene-1-sulfonyl chloride, dagenan chloride, 4-acetylamino benzenesulfonyl chloride, 4-acetamidophenylsulfonyl chloride, benzenesulfonyl chloride, 4-acetylamino, acetylsulfanilyl chloride, p-acetamidobenzenesulfonyl chloride, n-acetylsulphanilyl chloride, p-acetaminobenzenesulfonyl chloride | |
GRDXCFKBQWDAJH-UHFFFAOYSA-N | |
4-acetamidobenzenesulfonyl chloride | |
8481 | |
99% | |
N-Acetylsulfanilyl chloride, 99% |
98.5 | |
145.0°C to 148.0°C | |
>112°C | |
98.5% min. (Argentometry) | |
4-(CH3CONH)C6H4SO2Cl | |
14, 702 | |
15, 95 | |
Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylene dichloride,,alcohol,ether | |
CC(=O)NC1=CC=C(C=C1)S(Cl)(=O)=O | |
233.67 | |
233.67 | |
Granular Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-485-1
missing translation for 'rtecsNumber' : DB8837500
missing translation for 'tsca' : TSCA
RUO – Research Use Only