missing translation for 'onlineSavingsMsg'
Learn More
N-Allyl-N'-(2-hydroxyethyl)thiourea, 97%, Thermo Scientific Chemicals Product Code.: 11365149

N-Allyl-N'-(2-hydroxyethyl)thiourea, 97%, Thermo Scientific Chemicals

Product Code. 11365149
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11365149

Brand: Thermo Scientific Alfa Aesar A18161.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-81-7
Molecular Formula C6H12N2OS
Molecular Weight (g/mol) 160.235
MDL Number MFCD00002838
InChI Key VUVPNTYTOUGMDG-UHFFFAOYSA-N
Synonym 1-allyl-3-2-hydroxyethyl-2-thiourea, 1-allyl-3-2-hydroxyethyl thiourea, thiourea, n-2-hydroxyethyl-n'-2-propenyl, n-allyl-n'-2-hydroxyethyl thiourea, n-2-hydroxyethyl-n'-allylthiourea, n-2-hydroxyethyl-n'-2-propenylthiourea, n-allyl-n'-2-hydroxyethylthiourea, urea, 1-allyl-3-2-hydroxyethyl-2-thio, n-allyl-n'-beta-hydroxyethyl thioureaShow MoreShow Less
PubChem CID 2735273
IUPAC Name 1-(2-hydroxyethyl)-3-prop-2-enylthiourea
SMILES C=CCNC(=S)NCCO

Specifications

Melting Point 77°C to 79°C
Quantity 50 g
Formula Weight 160.24
Percent Purity 97%
Chemical Name or Material N-Allyl-N'-(2-hydroxyethyl)thiourea

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243650 Certificate TypeCertificate of Analysis Date26/09/2022 Catalog NumberA18161.30,A18161.18
Lot Number10241004 Certificate TypeCertificate of Analysis Date02/06/2022 Catalog NumberA18161.30,A18161.18
Lot Number10238182 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA18161.30,A18161.18
Lot Number10216645 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18161.30,A18161.18
Lot NumberC2340B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18161.30,A18161.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Allyl-N'-(2-hydroxyethyl)thiourea, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.