missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N(alpha)-Acetyl-L-lysine, 99%
Click to view available options
Quantity:
1 g
5 g
Description
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).
Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml--clear colorless solution)).
Notes
Protect from heat. Store away from oxidizing agents.
Nα-Acetyl-L-lysine is a biologically available N-terminal capped form of the proteinogenic α amino acid L-lysine. Nα-Acetyl-L-lysine is a substrate used to study and characterize lysyl oxidize(s).
Solubility
Soluble in water (miscible), and 80% acetic acid (50 mg/ml--clear colorless solution)).
Notes
Protect from heat. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 256°C to 258°C (decomposition) |
Color | White |
Solubility Information | Soluble in water (miscible),and 80% acetic acid (50mg/ml--clear colorless solution)). |
Formula Weight | 188.2 |
Percent Purity | 99% |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | N(alpha)-Acetyl-L-lysine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'