missing translation for 'onlineSavingsMsg'
Learn More

N(alpha)-Benzyloxycarbonyl-L-histidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7045014
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14997-58-1
C14H15N3O4
289.291
MFCD00065960
WCOJOHPAKJFUDF-LBPRGKRZSA-N
z-his-oh, n-cbz-l-histidine, cbz-his-oh, cbz-l-his-oh, nalpha-cbz-l-histidine, cbz-histidine, nalpha-carbobenzoxy-l-histidine, l-histidine, n-phenylmethoxy carbonyl, z-l-histidine, s-2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid
736313
(2S)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O
This item is not returnable. View return policy
14997-58-1
C14H15N3O4
289.291
MFCD00065960
WCOJOHPAKJFUDF-LBPRGKRZSA-N
z-his-oh, n-cbz-l-histidine, cbz-his-oh, cbz-l-his-oh, nalpha-cbz-l-histidine, cbz-histidine, nalpha-carbobenzoxy-l-histidine, l-histidine, n-phenylmethoxy carbonyl, z-l-histidine, s-2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid
736313
(2S)-3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽165°C (decomposition)
Beilstein 3561682
Formula Weight 289.29
Percent Purity ≥98%
Quantity 1 g
Optical Rotation −24° (c=6 in 6N HCl)
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-histidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'