missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-alpha-BOC-N-epsilon-(2-chloro-Z)-L-lysine, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Specifications
CAS | 54613-99-9 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Molecular Formula | C19H27ClN2O6 |
Quantity | 5 g |
Specific Rotation | -11 |
Synonym | boc-lys 2-cl-z-oh, boc-lys 2-cl-cbz-oh, n-boc-n'-2-chlorobenzyloxycarbonyl-l-lysine, n-boc-n-2-chloro-cbz-l-lysine, boc-l-lys 2-cl-z-oh, ambotzbaa1102, boc-lys clz-oh, pubchem12945, boc-lys cl-z-oh, boc-lys-2-cl-z-oh |
Show More |
Safety and Handling
Product Identifier
- N-alpha-BOC-N-epsilon-(2-Chloro-Z)-L-lysine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'