missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Amidinothiourea, 98+%, Thermo Scientific™
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.
Solubility
Soluble in water (70g/L).
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.
Solubility
Soluble in water (70g/L).
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 160°C to 168°C |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 1756620 |
Synonym | amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine |
Solubility Information | Soluble in water (70g/L). |
SMILES | C(=NC(=S)N)(N)N |
IUPAC Name | diaminomethylidenethiourea |
PubChem CID | 2724563 |
Formula Weight | 118.16 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'