missing translation for 'onlineSavingsMsg'
Learn More

N-Aminophthalimide, 94%

Product Code. 11408757
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
This item is not returnable. View return policy

Product Code. 11408757

Brand: Thermo Scientific Alfa Aesar A19914.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1875-48-5 | C8H6N2O2 | 162.148 g/mol

It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the aziridination of chiral N-enoyl sultams. It is also used in the synthesis of n-phthalimidoaziridines

Solubility
Solubility in hot methanol almost transparent.

Notes
Store away from oxidizing agents, strong bases and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1875-48-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005895
InChI Key KSILMCDYDAKOJD-UHFFFAOYSA-N
Synonym n-aminophthalimide, 2-aminoisoindoline-1,3-dione, 2-amino-1h-isoindole-1,3 2h-dione, phthalimide, n-amino, hydrazine, 1,1-phthaloyl, phthalhydrazide van, 1h-isoindole-1,3 2h-dione, 2-amino, n-aminophthalamide, phthalocyclohydrazide van, ccris 5349
PubChem CID 74645
IUPAC Name 2-aminoisoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)N

Specifications

Melting Point ∼195°C (decomposition)
Quantity 10 g
Beilstein 383756
Formula Weight 162.15
Percent Purity 94%
Chemical Name or Material N-Aminophthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.