Learn More
n-Amylamine, 99%, Thermo Scientific Chemicals
£16.70 - £133.00
Chemical Identifiers
CAS | 110-58-7 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.15 |
MDL Number | MFCD00008236 |
InChI Key | DPBLXKKOBLCELK-UHFFFAOYSA-N |
Synonym | amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine |
PubChem CID | 8060 |
ChEBI | CHEBI:74848 |
IUPAC Name | pentan-1-amine |
SMILES | CCCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521842
|
Thermo Scientific Acros
173940050 |
5 mL | Glass bottle |
£16.70
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10674852
|
Thermo Scientific Acros
173940500 |
50 mL | Glass bottle |
£41.10
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10770965
|
Thermo Scientific
173942500 |
250 mL | Glass bottle |
£133.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-58-7 | |
87.15 | |
DPBLXKKOBLCELK-UHFFFAOYSA-N | |
8060 | |
pentan-1-amine |
C5H13N | |
MFCD00008236 | |
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
CHEBI:74848 | |
CCCCCN |
Specifications
110-58-7 | |
100.0 | |
Colorless to Yellow | |
104.0°C | |
Authentic | |
C5H13N | |
1.4100 to 1.4120 | |
, 175 | |
0.75 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
CCCCCN | |
87.15 | |
CHEBI:74848 | |
87.15 | |
Liquid |
98.5 | |
-50.0°C | |
0.7500g/mL | |
4°C | |
98.5% min. (GC) | |
CH3(CH2)4NH2 | |
MFCD00008236 | |
15, 593 | |
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
DPBLXKKOBLCELK-UHFFFAOYSA-N | |
pentan-1-amine | |
8060 | |
1.2 mPa.s (20°C) | |
99% | |
n-Amylamine |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/sh
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-780-2
Recommended Storage : Product may darken during storage
- n-Amylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only