missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2,6-difluoro-N-isopropylbenzamide, 97%, Thermo Scientific™

Product Code. 15475568
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421262-76-1
Molecular Formula C17H17F2NO
Molecular Weight (g/mol) 289.326
MDL Number MFCD22683052
InChI Key WMGHZTPHABWXOF-UHFFFAOYSA-N
Synonym n-benzyl-2,6-difluoro-n-isopropylbenzamide
PubChem CID 97030377
IUPAC Name N-benzyl-2,6-difluoro-N-propan-2-ylbenzamide
SMILES CC(C)N(CC1=CC=CC=C1)C(=O)C2=C(C=CC=C2F)F

Specifications

Quantity 1 g
Formula Weight 289.32
Percent Purity 97%
Chemical Name or Material N-Benzyl-2,6-difluoro-N-isopropylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'