missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2-pyridineethylamine, Thermo Scientific™

£47.02 - £170.00

Chemical Identifiers

CAS 6312-25-0
Molecular Formula C14H16N2
Molecular Weight (g/mol) 212.296
MDL Number MFCD00089263
InChI Key WUDUNBALAGWKQB-UHFFFAOYSA-N
Synonym n-benzyl-2-pyridin-2-yl ethanamine, n-benzyl-2-pyridin-2-yl-ethanamine, benzyl 2-pyridin-2-yl ethyl amine, n-benzyl-2-pyridin-2-ylethylamine, benzyl 2-2-pyridyl ethyl amine, acmc-20ak6u, n-benzyl-2-pyridineethylamine, 2-2-benzylaminoethyl pyridine, benzyl-2-pyridin-2-yl-ethyl amine, n-phenylmethyl-2-pyridin-2-ylethanamineShow MoreShow Less
PubChem CID 238354
IUPAC Name N-benzyl-2-pyridin-2-ylethanamine
SMILES C1=CC=C(C=C1)CNCCC2=CC=CC=N2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11439390
SDS View Documents
Thermo Scientific Alfa Aesar
H50257.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11449390
SDS View Documents
Thermo Scientific Alfa Aesar
H50257.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzyl-2-pyridineethylamine, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.