missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-fluorobenzylamine, 97%, Thermo Scientific™

Product Code. 15442828
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55096-88-3
Molecular Formula C14H14FN
Molecular Weight (g/mol) 215.271
MDL Number MFCD00716882
InChI Key SXZSRGKJZKOZRP-UHFFFAOYSA-N
Synonym n-benzyl-1-4-fluorophenyl methanamine, benzyl-4-fluoro-benzyl-amine, benzyl-4-fluorobenzyl amine, benzyl 4-fluorophenyl methyl amine, benzyl 4-fluorobenzyl amine, 4-fluorophenyl methyl benzylamine, n-benzyl-n-4-fluorobenzyl amine, benzenemethanamine,4-fluoro-n-phenylmethyl, n-4-fluorophenyl methyl-1-phenylmethanamine
PubChem CID 784761
IUPAC Name N-[(4-fluorophenyl)methyl]-1-phenylmethanamine
SMILES C1=CC=C(C=C1)CNCC2=CC=C(C=C2)F

Specifications

Quantity 250 mg
Formula Weight 215.27
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-fluorobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'