missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-4-methoxy-N-phenylbenzamide, 97%, Thermo Scientific™

Product Code. 15485338
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111735-27-4
Molecular Formula C21H19NO2
Molecular Weight (g/mol) 317.39
MDL Number MFCD22683043
InChI Key BLUMCNOTEFEZGV-UHFFFAOYSA-N
Synonym benzamide, 4-methoxy-n-phenyl-n-phenylmethyl, acmc-20mep6
PubChem CID 13736433
IUPAC Name N-benzyl-4-methoxy-N-phenylbenzamide
SMILES COC1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 317.39
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-methoxy-N-phenylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'