missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-beta-alanine ethyl ester, 95%, Thermo Scientific™

Product Code. 11318596
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11318596

Brand: Thermo Scientific Alfa Aesar L17652.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23583-21-3
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD00085541
InChI Key HCTJHQFFNDLDPF-UHFFFAOYSA-N
Synonym ethyl 3-benzylamino propanoate, ethyl 3-benzylamino propionate, ethyl n-benzyl-beta-alaninate, n-benzyl-beta-alanine ethyl ester, 3-benzylamino-propionic acid ethyl ester, 3-benzylaminopropionic acid ethyl ester, n-benzyl-3-aminopropionic acid ethyl ester, ethyl n-benzyl-, a-alaninate, ethyl3-benzylamino propanoate
PubChem CID 90196
IUPAC Name ethyl 3-(benzylamino)propanoate
SMILES CCOC(=O)CCNCC1=CC=CC=C1

Specifications

Boiling Point 150°C (0.2mmHg)
Linear Formula C6H5CH2NHCH2CH2CO2CH2CH3
Refractive Index 1.505
Density 1.03
Quantity 1 g
Formula Weight 207.27
Percent Purity 98%
Chemical Name or Material N-Benzyl-beta-alanine ethyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.