missing translation for 'onlineSavingsMsg'
Learn More
N-Benzyl-p-toluenesulfonamide, Thermo Scientific Chemicals Product Code.: 15426639

N-Benzyl-p-toluenesulfonamide, Thermo Scientific Chemicals

Product Code. 15426639
5 mg
Click to view available options
Quantity:
5 mg
10 mg
This item is not returnable. View return policy

Product Code. 15426639

Brand: Thermo Scientific Alfa Aesar J64910.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A potent inhibitor of calcium-stimulated myosin S1 ATPase

A potent inhibitor of calcium-stimulated myosin S1 ATPase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-37-0
Molecular Formula C14H15NO2S
Molecular Weight (g/mol) 261.339
MDL Number MFCD00159328
InChI Key WTHKAJZQYNKTCJ-UHFFFAOYSA-N
Synonym n-benzyl-p-toluenesulfonamide, n-tosylbenzylamine, p-toluenesulfonamide, n-benzyl, n-benzyl-p-toluenesulphonamide, n-benzyl-4-methyl-benzenesulfonamide, n-benzyl-4-methylbenzene-1-sulfonamide, n-benzyl-p-toluene sulphonamide, benzenesulfonamide, 4-methyl-n-phenylmethyl, 4-methyl-n-phenylmethyl benzenesulfonamide, 4-methyl-n-phenylmethyl-benzenesulfonamid
PubChem CID 95801
IUPAC Name N-benzyl-4-methylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)NCC2=CC=CC=C2

Specifications

Melting Point 115°C
Color White
Quantity 5 mg
Sensitivity Light sensitive
Formula Weight 261.34
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-p-toluenesulfonamide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14J018 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberJ64910.#0,J64910.MA
Lot NumberU25J019 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ64910.#0,J64910.MA
Lot NumberR18J005 Certificate TypeCertificate of Analysis Date25/05/2023 Catalog NumberJ64910.#0,J64910.MA
Lot NumberX05I015 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog NumberJ64910.#0,J64910.MA
Lot NumberN21I053 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberJ64910.#0,J64910.MA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzyl-p-toluenesulfonamide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.