missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzylbenzamide, 99+%

Product Code. 11424873
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy
Applications
N-Benzylbenzamide is a convenient precursor to α-substituted benzyl amines and an indicator for the titration of butyl lithium and other lithium bases.

Solubility
Soluble in Acetone (25 mg/ml).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1485-70-7
Melting Point 104°C to 108°C
Molecular Formula C14H13NO
MDL Number MFCD00003070
Quantity 25 g
Beilstein 2211315
Synonym benzamide, n-phenylmethyl, n-benzyl-benzamide, phenyl-n-benzylcarboxamide, benzoylbenzylamine, n∼1∼-benzylbenzamide, benzamide, n-benzyl, n-phenylmethyl benzamide, acmc-1bx9y, n-benzylbenzamide, lkqucicfthbfal-uhfffaoysa
InChI Key LKQUCICFTHBFAL-UHFFFAOYSA-N
SMILES O=C(NCC1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name N-benzylbenzamide
Molecular Weight (g/mol) 211.26
PubChem CID 73878
Formula Weight 211.26
Percent Purity ≥99%
Chemical Name or Material N-Benzylbenzamide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'