missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzylmethylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
1000 mL
Description
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
Solubility
Soluble in water (65 g/L at 20°C).
Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.
Solubility
Soluble in water (65 g/L at 20°C).
Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
Specifications
Specifications
Melting Point | -55°C |
Density | 0.94 |
pH | 10 |
Boiling Point | 184°C to 188°C |
Flash Point | 77°C (170°F) |
Odor | Amine-like |
Linear Formula | C6H5CH2NHCH3 |
Refractive Index | 1.523 |
Quantity | 1000 mL |
UN Number | UN2922 |
Show More |
Safety and Handling
Product Identifier
- N-Benzylmethylamine
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : N-Benzylmethylamine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction