missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylmethylamine, 97%, Thermo Scientific Chemicals

£20.90 - £69.35

Chemical Identifiers

CAS 103-67-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008289
InChI Key RIWRFSMVIUAEBX-UHFFFAOYSA-N
Synonym n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
PubChem CID 7669
IUPAC Name N-methyl-1-phenylmethanamine
SMILES CNCC1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11468760
View Documents
Thermo Scientific Alfa Aesar
A13746.AE
100 mL
£20.90
100mL
Please to purchase this item. Need a web account? Register with us today!
11454784
View Documents
Thermo Scientific Alfa Aesar
A13746.AK
250 mL
£26.53
250mL
Please to purchase this item. Need a web account? Register with us today!
11444784
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13746.AU
1000 mL
£69.35
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

Solubility
Soluble in water (65 g/L at 20°C).

Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
Specifications

Chemical Identifiers

103-67-3
121.18
RIWRFSMVIUAEBX-UHFFFAOYSA-N
7669
CNCC1=CC=CC=C1
C8H11N
MFCD00008289
n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
N-methyl-1-phenylmethanamine

Specifications

103-67-3
0.94
184°C to 188°C
Amine-like
C6H5CH2NHCH3
MFCD00008289
606221
n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
RIWRFSMVIUAEBX-UHFFFAOYSA-N
N-methyl-1-phenylmethanamine
7669
97%
-55°C
10
77°C (170°F)
C8H11N
1.523
UN2922
Air Sensitive
Soluble in water (65g/L at 20°C).
CNCC1=CC=CC=C1
121.18
121.18
N-Benzylmethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H301-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 203-133-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • N-Benzylmethylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : N-Benzylmethylamine

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.