missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, 97%, Thermo Scientific™

Product Code. 15438126
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17400-34-9
Molecular Formula C11H17ClN2O2
Molecular Weight (g/mol) 244.72
MDL Number MFCD00270153
InChI Key XKMBTMXQMDLSRB-UHFFFAOYSA-N
Synonym n-cbz-1,3-diaminopropane hydrochloride, benzyl 3-aminopropyl carbamate hydrochloride, benzyl n-3-aminopropyl carbamate hydrochloride, n-cbz-1,3-diaminopropane-hcl, n-z-1,3-propanediamine hydrochloride, n-carbobenzoxy-1,3-diaminopropane hydrochloride, n-carbobenzyloxy-1,3-diaminopropane hydrochloride, n-3-aminopropyl carbamic acid benzyl ester hydrochloride, n-1-z-1,3-diaminopropane hcl, n-z-1,3-diaminopropane hydrochloride
PubChem CID 13196227
IUPAC Name benzyl N-(3-aminopropyl)carbamate;hydrochloride
SMILES [H+].[Cl-].NCCCNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 185°C to 189°C
Color White
Quantity 1 g
Beilstein 4725495
Sensitivity Hygroscopic
Formula Weight 244.72
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'