missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, 97%, Thermo Scientific Chemicals

Catalog Number p-7036758
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
65706-99-2
C20H21NO6
371.389
MFCD00069647
VWHKODOUMSMUAF-QGZVFWFLSA-N
z-d-glu-obzl, cbz-d-glu-obzl, n-cbz-d-glutamic acid alpha-benzyl ester, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, cbz-d-glu-och2ph, pubchem14966, cbz-d-glu oh-obzl
7018162
(4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
65706-99-2
C20H21NO6
371.389
MFCD00069647
VWHKODOUMSMUAF-QGZVFWFLSA-N
z-d-glu-obzl, cbz-d-glu-obzl, n-cbz-d-glutamic acid alpha-benzyl ester, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, cbz-d-glu-och2ph, pubchem14966, cbz-d-glu oh-obzl
7018162
(4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, is used as a reagent in the synthesis of acyl peptides that act as immunostimulants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester, is used as a reagent in the synthesis of acyl peptides that act as immunostimulants.

Solubility
Soluble in methanol and acetic acid.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 5305848
Solubility Information Soluble in methanol and acetic acid.
Formula Weight 371.39
Percent Purity 97%
Chemical Name or Material N-Benzyloxycarbonyl-D-glutamic acid 1-benzyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'