missing translation for 'onlineSavingsMsg'
Learn More
N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals Product Code.: 15447266

N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15447266
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15447266

Brand: Thermo Scientific Alfa Aesar H62306.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4779-31-1
Molecular Formula C19H19NO6
Molecular Weight (g/mol) 357.36
MDL Number MFCD00038594
InChI Key UYOZWZKJQRBZRH-UHFFFAOYNA-N
Synonym z-asp-obzl, n-carbobenzyloxy-l-aspartic acid 1-benzyl ester, 1-benzyl n-carbobenzyloxy-l-aspartate, 3s-4-benzyloxy-3-benzyloxy carbonyl amino-4-oxobutanoic acid, 3s-4-oxo-4-phenylmethoxy-3-phenylmethoxycarbonylamino butanoic acid, z-asp-obn, cbz-l-asp-obn, benzyloxycarbonyl-l-aspartic-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-benzyl ester, 2-benzyl-oxycarbonylamino-succinic acid 1-benzyl ester
PubChem CID 6994509
IUPAC Name 4-(benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
SMILES OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 357.36
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX09G033 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62306.06,H62306.03,H62306.14
Lot NumberN21D018 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62306.06,H62306.03,H62306.14
Lot NumberZ27C011 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62306.06,H62306.03,H62306.14
Lot NumberM15E006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62306.06,H62306.03,H62306.14
Lot Number5004F17V Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62306.06,H62306.03,H62306.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.