missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15498596
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5891-45-2
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.37
MDL Number MFCD00077016
InChI Key VJECGKAFPHEJQS-UHFFFAOYNA-N
Synonym z-glu-otbu, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18089, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, a-tert.butyl ester
PubChem CID 6994036
IUPAC Name 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1

Specifications

Formula Weight 337.37
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'