missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Product Code. 10124650
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13139-17-8
Molecular Formula C12H11NO5
Molecular Weight (g/mol) 249.22
MDL Number MFCD00005513
InChI Key MJSHDCCLFGOEIK-UHFFFAOYSA-N
Synonym n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide
PubChem CID 83172
IUPAC Name benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 77°C to 82°C
Color White
Loss on Drying 0.25% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information (10% in acetone) Clear colorless to almost colorless
Formula Weight 249.22
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-(Benzyloxycarbonyloxy)succinimide
Show More Show Less
Product Identifier
  • N-(Benzyloxycarbonyloxy)succinimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'