missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyloxy)succinimide, 99%, Thermo Scientific Chemicals

Product Code. 11490123
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

Solubility
Soluble in chloroform, methanol, acetone (0.1 g/mL).

Notes
Moisture sensitive. Storage temperature 2-8°C. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 13139-17-8
Molecular Formula C12H11NO5
Molecular Weight (g/mol) 249.222
MDL Number MFCD00005513
InChI Key MJSHDCCLFGOEIK-UHFFFAOYSA-N
Synonym n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide
PubChem CID 83172
IUPAC Name benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2

Specifica

Melting Point 77°C to 82°C
Quantity 100 g
Beilstein 1387927
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methanol,acetone (0.1g/mL).
Formula Weight 249.23
Percent Purity 99%
Chemical Name or Material N- (Benzyloxycarbonyloxy)succinimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'