missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 25 g |
Formula Weight | 215.29 |
Percent Purity | 97% |
Chemical Name or Material | 8-Benzyl-8-azabicyclo[3.2.1]octan-3-one |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-Benzyltropinone, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction