missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,4-diaminocyclohexane, 95%, Thermo Scientific Chemicals

Product Code. 15442359
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195314-59-1
Molecular Formula C11H23N2O2
Molecular Weight (g/mol) 215.32
MDL Number MFCD01076211,MFCD03001719,MFCD03844604
InChI Key FEYLUKDSKVSMSZ-UHFFFAOYSA-O
Synonym trans-n-boc-1,4-cyclohexanediamine, n-boc-trans-1,4-cyclohexanediamine, 1-n-boc-cis-1,4-cyclohexyldiamine, tert-butyl 4-aminocyclohexyl carbamate, n-boc-1,4-cyclohexanediamine, tert-butyl n-4-aminocyclohexyl carbamate, tert-butyl cis-4-aminocyclohexyl carbamate, 1-boc-amino-1,4-cyclohexanediamine, tert-butyl 4-aminocyclohexylcarbamate, cis-n-boc-1,4-cyclohexanediamine
PubChem CID 2756050
IUPAC Name tert-butyl N-(4-aminocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC([NH3+])CC1

Specifications

Color White
Quantity 5 g
UN Number UN3259
Beilstein 13428745
Formula Weight 214.31
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-1,4-diaminocyclohexane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'