missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-2-bromo-D-phenylalanine, 95%, Thermo Scientific™
£39.80 - £375.00
Chemical Identifiers
CAS | 261360-76-3 |
---|---|
Molecular Formula | C14H18BrNO4 |
Molecular Weight (g/mol) | 344.205 |
MDL Number | MFCD01317703 |
InChI Key | XDJSTMCSOXSTGZ-LLVKDONJSA-N |
Synonym | boc-2-bromo-d-phenylalanine, boc-d-phe 2-br-oh, n-boc-2-bromo-d-phenylalanine, boc-d-2-bromophenylalanine, r-n-boc-2-bromophenylalanine, boc-d-2-bromophe, r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-2-br-phe-oh, 2r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, n-boc-d-2-bromophenylalanine |
PubChem CID | 7006627 |
IUPAC Name | (2R)-3-(2-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Br)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsN-Boc-2-bromo-D-phenylalanine is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Notes
Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
261360-76-3 | |
344.205 | |
XDJSTMCSOXSTGZ-LLVKDONJSA-N | |
7006627 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Br)C(=O)O |
C14H18BrNO4 | |
MFCD01317703 | |
boc-2-bromo-d-phenylalanine, boc-d-phe 2-br-oh, n-boc-2-bromo-d-phenylalanine, boc-d-2-bromophenylalanine, r-n-boc-2-bromophenylalanine, boc-d-2-bromophe, r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-2-br-phe-oh, 2r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, n-boc-d-2-bromophenylalanine | |
(2R)-3-(2-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
261360-76-3 | |
C14H18BrNO4 | |
boc-2-bromo-d-phenylalanine, boc-d-phe 2-br-oh, n-boc-2-bromo-d-phenylalanine, boc-d-2-bromophenylalanine, r-n-boc-2-bromophenylalanine, boc-d-2-bromophe, r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-2-br-phe-oh, 2r-3-2-bromophenyl-2-tert-butoxycarbonyl amino propanoic acid, n-boc-d-2-bromophenylalanine | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1Br)C(=O)O | |
344.205 | |
344.2 | |
Powder |
White | |
MFCD01317703 | |
XDJSTMCSOXSTGZ-LLVKDONJSA-N | |
(2R)-3-(2-bromophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
7006627 | |
95% | |
N-Boc-2-bromo-D-phenylalanine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures