missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-3-(4-biphenylyl)-D-alanine, 95%, Thermo Scientific Chemicals
POA - £269.00
Chemical Identifiers
CAS | 128779-47-5 |
---|---|
Molecular Formula | C20H23NO4 |
Molecular Weight (g/mol) | 341.41 |
MDL Number | MFCD00191185 |
InChI Key | NBVVKAUSAGHTSU-UHFFFAOYNA-N |
Synonym | boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh |
PubChem CID | 7019133 |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15450219
|
Thermo Scientific Alfa Aesar
H63494.MD |
250 mg | |||||||
15430219
|
Thermo Scientific Alfa Aesar
H63494.03 |
1 g | |||||||
15440219
|
Thermo Scientific Alfa Aesar
H63494.06 |
5 g |
£269.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
128779-47-5 | |
341.41 | |
NBVVKAUSAGHTSU-UHFFFAOYNA-N | |
7019133 |
C20H23NO4 | |
MFCD00191185 | |
boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O |
Specifications
128779-47-5 | |
MFCD00191185 | |
NBVVKAUSAGHTSU-UHFFFAOYNA-N | |
3-{[1,1'-biphenyl]-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid | |
7019133 | |
95% | |
N-Boc-3-(4-biphenylyl)-D-alanine |
C20H23NO4 | |
boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh | |
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O | |
341.41 | |
341.41 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only