missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-bromoaniline, 97%, Thermo Scientific Chemicals

Product Code. 15481398
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131818-17-2
Molecular Formula C11H14BrNO2
Molecular Weight (g/mol) 272.14
MDL Number MFCD01006612
InChI Key VLGPDTPSKUUHKR-UHFFFAOYSA-N
Synonym tert-butyl 4-bromophenyl carbamate, tert-butyl n-4-bromophenyl carbamate, tert-butyl 4-bromophenylcarbamate, n-boc-4-bromoaniline, n-tert-butoxycarbonyl-4-bromoaniline, metronidazolebenzoate, n-boc 4-bromoaniline, 4-bromo-n-boc aniline, acmc-1c4z5, t-butyl 4-bromophenylcarbamate
PubChem CID 2773608
IUPAC Name tert-butyl N-(4-bromophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(Br)C=C1

Specifications

Melting Point 103°C to 106°C
Quantity 1 g
Formula Weight 272.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-4-bromoaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'