missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-iodoaniline, 95%, Thermo Scientific™

Product Code. 15483787
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159217-89-7
Molecular Formula C11H14INO2
Molecular Weight (g/mol) 319.14
MDL Number MFCD03490497
InChI Key CGALRQWBJFDAKN-UHFFFAOYSA-N
Synonym tert-butyl n-4-iodophenyl carbamate, n-boc-4-iodoaniline, tert-butyl 4-iodophenyl carbamate, tert-butyl 4-iodophenylcarbamate, 4-iodo-phenyl-carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-4-iodoaniline, carbamic acid, 4-iodophenyl-, 1,1-dimethylethyl ester, carbamic acid, n-4-iodophenyl-, 1,1-dimethylethyl ester, n-boc 4-iodoaniline, acmc-1c8b7
PubChem CID 2773612
IUPAC Name tert-butyl N-(4-iodophenyl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(I)C=C1

Specifications

Melting Point 142°C to 144°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 319.14
Percent Purity 95%
Chemical Name or Material N-Boc-4-iodoaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'