missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-nitro-D-phenylalanine, 98%, Thermo Scientific™

£40.10 - £141.00

Chemical Identifiers

CAS 61280-75-9
Molecular Formula C14H18N2O6
Molecular Weight (g/mol) 310.31
MDL Number MFCD00069880
InChI Key XBQADBXCNQPHHY-UHFFFAOYNA-N
Synonym boc-4-nitro-d-phenylalanine, boc-d-phe 4-no2-oh, boc-d-4-nitrophe, boc-d-4-no2-phe-oh, r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-d-phenylalanine, boc-d-4-nitrophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-nitro, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-4-nitrophenyl propanoic acid
PubChem CID 2755957
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-(4-nitrophenyl)propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15458228
SDS View Documents
Thermo Scientific Alfa Aesar
H61669.03
1 g N/A N/A N/A
15468228
SDS View Documents
Thermo Scientific Alfa Aesar
H61669.06
5 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-4-nitro-D-phenylalanine, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.