missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Catalog Number 15493777
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
102195-80-2
C11H17NO5
243.259
MFCD01861778
UPBHYYJZVWZCOZ-QMMMGPOBSA-N
boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
10988485
1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC
This item is not returnable. View return policy
102195-80-2
C11H17NO5
243.259
MFCD01861778
UPBHYYJZVWZCOZ-QMMMGPOBSA-N
boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
10988485
1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 40°C
Quantity 250 mg
Formula Weight 243.26
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline methyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'