missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-4-Piperidone, 99%, Thermo Scientific Chemicals

Product Code. 10584911
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79099-07-3
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00151800
InChI Key ROUYFJUVMYHXFJ-UHFFFAOYSA-N
Synonym 1-boc-4-piperidone, n-boc-4-piperidone, n-tert-butoxycarbonyl-4-piperidone, 1-boc-4-piperidinone, boc-4-piperidone, 1-tert-butoxycarbonyl-4-piperidone, n-boc-4-piperidinone, boc-piperidone, 1-t-boc-4-piperidone
PubChem CID 735900
IUPAC Name tert-butyl 4-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 74.0°C to 75.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 199.25
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-4-Piperidone
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-4-Piperidone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'