missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-BOC-D-Phenylalaninal, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Melting Point | 85.0°C to 89.0°C |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Tan or White |
Assay Percent Range | 96% min. (GC) |
Infrared Spectrum | Authentic |
Specific Rotation | + 45.00 |
Formula Weight | 249.31 |
Percent Purity | 97% |
Quantity | 250 mg |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'