missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-valine, 98+%, Thermo Scientific Chemicals

Product Code. 11357276
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22838-58-0
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00038282
InChI Key SZXBQTSZISFIAO-SSDOTTSWSA-N
Synonym boc-d-valine, boc-d-val-oh, n-boc-d-valine, n-tert-butoxycarbonyl-d-valine, tert-butoxycarbonyl-d-valine, r-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, d-valine, n-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-valin, r-2-tert-butoxycarbonylamino-3-methylbutanoic acid, n-alpha-t-butyloxycarbonyl-d-valine
PubChem CID 637605
IUPAC Name (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 76°C to 80°C
Quantity 5 g
Beilstein 2050408
Optical Rotation +6.3° (c=1 in Acetic Acid)
Formula Weight 217.27
Percent Purity ≥98%
Chemical Name or Material N-Boc-D-valine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'