missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-glycylglycine, 97%, Thermo Scientific Chemicals
£92.70 - £315.00
Chemical Identifiers
CAS | 31972-52-8 |
---|---|
Molecular Formula | C9H16N2O5 |
Molecular Weight (g/mol) | 232.236 |
MDL Number | MFCD00038558 |
InChI Key | HWBAHOVOSOAFLE-UHFFFAOYSA-N |
Synonym | boc-gly-gly-oh, boc-gly-gly, n-tert-butoxycarbonyl glycylglycine, n-2-n-boc-amino-acetyl-glycine, boc-glycylglycine, 2-2-tert-butoxycarbonyl amino acetamido acetic acid, 2-tert-butoxycarbonyl amino acetamido acetic acid, 2-2-tert-butoxycarbonylamino acetamido acetic acid, 2-2-tert-butoxy carbonyl amino acetamido acetic acid, t-boc-glygly |
PubChem CID | 333466 |
IUPAC Name | 2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid |
SMILES | CC(C)(C)OC(=O)NCC(=O)NCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15469338
|
Thermo Scientific Alfa Aesar
H62889.06 |
5 g |
£92.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479338
|
Thermo Scientific Alfa Aesar
H62889.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
31972-52-8 | |
232.236 | |
HWBAHOVOSOAFLE-UHFFFAOYSA-N | |
333466 | |
CC(C)(C)OC(=O)NCC(=O)NCC(=O)O |
C9H16N2O5 | |
MFCD00038558 | |
boc-gly-gly-oh, boc-gly-gly, n-tert-butoxycarbonyl glycylglycine, n-2-n-boc-amino-acetyl-glycine, boc-glycylglycine, 2-2-tert-butoxycarbonyl amino acetamido acetic acid, 2-tert-butoxycarbonyl amino acetamido acetic acid, 2-2-tert-butoxycarbonylamino acetamido acetic acid, 2-2-tert-butoxy carbonyl amino acetamido acetic acid, t-boc-glygly | |
2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid |
Specifications
31972-52-8 | |
C9H16N2O5 | |
MFCD00038558 | |
HWBAHOVOSOAFLE-UHFFFAOYSA-N | |
2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]acetic acid | |
333466 | |
97% | |
N-Boc-glycylglycine |
White | |
(CH3)3COCONHCH2CONHCH2CO2H | |
boc-gly-gly-oh, boc-gly-gly, n-tert-butoxycarbonyl glycylglycine, n-2-n-boc-amino-acetyl-glycine, boc-glycylglycine, 2-2-tert-butoxycarbonyl amino acetamido acetic acid, 2-tert-butoxycarbonyl amino acetamido acetic acid, 2-2-tert-butoxycarbonylamino acetamido acetic acid, 2-2-tert-butoxy carbonyl amino acetamido acetic acid, t-boc-glygly | |
CC(C)(C)OC(=O)NCC(=O)NCC(=O)O | |
232.236 | |
232.23 | |
Powder |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only