missing translation for 'onlineSavingsMsg'
Learn More
N-Boc-hexahydro-1H-azepin-4-one, 98%, Thermo Scientific Chemicals Product Code.: 15411388

N-Boc-hexahydro-1H-azepin-4-one, 98%, Thermo Scientific Chemicals

Product Code. 15411388
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15411388

Brand: Thermo Scientific Alfa Aesar H55214.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.

Solubility
Soluble in Chloroform, Dichloromethane, Ethanol and Ethyl Ether.

Notes
Air sensitive. Keep under 2-8°C. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188975-88-4
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD03788435
InChI Key PMLBUVZPRKXMOX-UHFFFAOYSA-N
Synonym n-boc-hexahydro-1h-azepin-4-one, 1-boc-hexahydro-1h-azepin-4-one, 4-oxoazepane-1-carboxylic acid tert-butyl ester, 1-boc-homopiperazin-4-one, 1-boc-4-azepanone, n-boc-hexahydro-1h-azepine-4-one, tert-butyl hexahydro-4-oxo-1h-azepine-1-carboxylate, 1-tert-butoxycarbonyl-4-oxoazepane, 4-oxo-azepane-1-carboxylic acid tert-butyl ester, 1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
PubChem CID 1512679
IUPAC Name tert-butyl 4-oxoazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)CC1

Specifications

Refractive Index 1.467
Quantity 5 g
Sensitivity Air sensitive
Solubility Information Soluble in Chloroform,Dichloromethane,Ethanol and Ethyl Ether.
Formula Weight 213.28
Percent Purity 98%
Chemical Name or Material N-Boc-hexahydro-1H-azepin-4-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5004R18N Certificate TypeCertificate of Analysis Date26/07/2024 Catalog NumberH55214.03,H55214.06
Lot Number5003K26P Certificate TypeCertificate of Analysis Date26/07/2024 Catalog NumberH55214.03,H55214.06
Lot NumberBI220554 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55214.03,H55214.06
Lot Number5004R18N Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55214.03,H55214.06
Lot Number5003K26P Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55214.03,H55214.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-hexahydro-1H-azepin-4-one, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.