missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-alaninamide, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
N-Boc-L-alaninamide is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-alaninamide is used as pharmaceutical intermediate.
Solubility
Soluble in water or 1% acetic acid.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
N-Boc-L-alaninamide is used as pharmaceutical intermediate.
Solubility
Soluble in water or 1% acetic acid.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 g |
Solubility Information | Soluble in water or 1% acetic acid. |
IUPAC Name | tert-butyl N-[(2S)-1-amino-1-oxopropan-2-yl]carbamate |
Formula Weight | 188.22 |
Percent Purity | 96% |
Physical Form | Solid |
Chemical Name or Material | N-Boc-L-alaninamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'