missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-aspartic acid 1-benzyl ester, 98%, Thermo Scientific Chemicals

Product Code. 11406006
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Boc-L-aspartic acid 1-benzyl ester is used as a local anesthetic and also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-aspartic acid 1-benzyl ester is used as a local anesthetic and also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30925-18-9
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00065563
InChI Key LDRWTKQWSXGSTM-LBPRGKRZSA-N
Synonym boc-asp-obzl, boc-l-aspartic acid 1-benzyl ester, 1-benzyl n-boc-l-aspartate, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, commericial, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, pubchem12927, boc-l-asp h-obn
PubChem CID 7017887
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 95°C to 97°C
Quantity 5 g
Solubility Information Soluble in dichloromethane
Formula Weight 323.35
Percent Purity 99%
Chemical Name or Material N-Boc-L-aspartic acid 1-benzyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'