missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid 5-tert-butyl ester, 99%, Thermo Scientific Chemicals

Catalog Number p-7026359
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13726-84-6
C14H25NO6
303.355
MFCD00038257
YGSRAYJBEREVRB-VIFPVBQESA-N
boc-glu otbu-oh, boc-l-glu otbu-oh, s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-l-glutamic acid 5-tert-butyl ester, n-boc-l-glutamic acid 5-tert-butyl ester, n-tert-butoxycarbonyl-l-glutamic acid gamma-tert-butyl ester, 2s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-glu otbu, ambotzbaa1297, pubchem18923
6993432
(2S)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
13726-84-6
C14H25NO6
303.355
MFCD00038257
YGSRAYJBEREVRB-VIFPVBQESA-N
boc-glu otbu-oh, boc-l-glu otbu-oh, s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-l-glutamic acid 5-tert-butyl ester, n-boc-l-glutamic acid 5-tert-butyl ester, n-tert-butoxycarbonyl-l-glutamic acid gamma-tert-butyl ester, 2s-5-tert-butoxy-2-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-glu otbu, ambotzbaa1297, pubchem18923
6993432
(2S)-5-[(2-methylpropan-2-yl)oxy]-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
CC(C)(C)OC(=O)CCC(C(=O)O)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 110°C to 112°C
Quantity 1 g
Formula Weight 303.36
Percent Purity 99%
Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'