missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-phenylalanine methyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15429568
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51987-73-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00076977
InChI Key SDSWSVBXRBXPRL-UHFFFAOYNA-N
Synonym boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507
PubChem CID 9882138
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
SMILES COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 36°C to 40°C
Color White
Beilstein 3061910
Quantity 25 g
Flash Point 113°C (235°F)
Formula Weight 379.33
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-L-phenylalanine methyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Boc-L-phenylalanine methyl ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.