missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-valine, 98+%, Thermo Scientific Chemicals
£23.90 - £91.20
Chemical Identifiers
CAS | 13734-41-3 |
---|---|
Molecular Formula | C10H19NO4 |
Molecular Weight (g/mol) | 217.27 |
MDL Number | MFCD00065605 |
InChI Key | SZXBQTSZISFIAO-ZETCQYMHSA-N |
Synonym | boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine |
PubChem CID | 83693 |
IUPAC Name | (2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
SMILES | CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458616
|
Thermo Scientific Alfa Aesar
A16007.06 |
5 g |
£23.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468616
|
Thermo Scientific Alfa Aesar
A16007.14 |
25 g |
£91.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Boc-L-valine is used in the multi-peptide synthesis and serves as the amino acid protection monomer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-valine is used in the multi-peptide synthesis and serves as the amino acid protection monomer.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
13734-41-3 | |
217.27 | |
SZXBQTSZISFIAO-ZETCQYMHSA-N | |
83693 | |
CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O |
C10H19NO4 | |
MFCD00065605 | |
boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine | |
(2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
13734-41-3 | |
C10H19NO4 | |
1711290 | |
Insoluble in water. | |
−7.5° (c=1 in Acetic Acid) | |
(2S)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
83693 | |
≥98% |
77°C to 80°C | |
MFCD00065605 | |
boc-val-oh, boc-l-valine, n-tert-butoxycarbonyl-l-valine, tert-butoxycarbonylvaline, boc-l-val-oh, s-2-boc-amino-3-methylbutyric acid, s-2-tert-butoxycarbonyl amino-3-methylbutanoic acid, n-boc-l-valine, n-tert-butyloxycarbonyl-l-valine | |
SZXBQTSZISFIAO-ZETCQYMHSA-N | |
CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
217.27 | |
217.27 | |
N-Boc-L-valine |
Safety and Handling
missing translation for 'einecsNumber' : 237-307-6
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only