missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-N(im)-tosyl-L-histidine, 98%, Thermo Scientific™

Product Code. 11925771
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35899-43-5
Molecular Formula C18H23N3O6S
Molecular Weight (g/mol) 409.46
MDL Number MFCD00065967
InChI Key DCLJSEPKYJSEHW-UHFFFAOYNA-N
Synonym boc-his tos-oh, boc-l-his tos-oh, boc-his tos, boc-l-histidine tosyl, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, boc-his-tos, ambotzbaa1153, pubchem7363, boc-his-tos-oh
PubChem CID 6455010
IUPAC Name (2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=NC(CC(NC(=O)OC(C)(C)C)C(O)=O)=C1

Specifications

Melting Point 117.0°C to 118.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Specific Rotation + 17.30
Formula Weight 409.46
Percent Purity 98%
Chemical Name or Material N-α-BOC-N(im)-tosyl-L-histidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'