missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-methyl-L-threonine, 95%, Thermo Scientific™

Product Code. 15480599
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48068-25-3
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00076990
InChI Key VWSUOKFUIPMDDX-RQJHMYQMSA-N
Synonym boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid
PubChem CID 15460346
IUPAC Name (2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CO[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Formula Weight 233.26
Percent Purity 95%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Boc-O-methyl-L-threonine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'